Gene Transcripts

Gene Symbol
SLC22A14
Entrez Gene ID
9389
Gene Name
solute carrier family 22 member 14
Gene Aliases
OCTL2, OCTL4, ORCTL4
Chromosome Location
Chr. 3: 38281662 - 38318575 on Build GRCh38
UniGene ID
Hs.165559
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320033.1 NP_001306962.1 2 126
NM_004803.4 NP_004794.2 1 171
XM_005265585.4 XP_005265642.1 2 948
XM_006713416.3 XP_006713479.2 2 872
XM_006713417.3 XP_006713480.2 2 872
XM_006713418.3 XP_006713481.2 2 872
XM_011534245.2 XP_011532547.1 2 960
XM_011534247.2 XP_011532549.1 2 872
GenBank mRNA AB011082.1 - 1 170
AK130497.1 - 1 934
AK315317.1 - 2 124
BC075070.2 - 1 115 30915378
BC075071.2 - 1 115 30915591
BC126482.1 - 1 152 8992198
BC130543.1 - 1 152 40146574
BC144079.1 - 1 152
DB024161.1 - 2 128

View Full Product Details