Gene Transcripts

Gene Symbol
DDX23
Entrez Gene ID
9416
Gene Name
DEAD-box helicase 23
Gene Aliases
PRPF28, SNRNP100, U5-100K, U5-100KD, prp28
Chromosome Location
Chr. 12: 48829756 - 48852174 on Build GRCh38
UniGene ID
Hs.130098
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004818.2 NP_004809.2 3 342
GenBank mRNA AF026402.1 - 3 290
AK128428.1 - 3 331
AK292921.1 - 3 331
AK294877.1 - 3 309
AK312379.1 - 3 311
BC002366.2 - 3 301 2820898
CD709611.1 - 3 374

View Full Product Details