Gene Transcripts

Gene Symbol
Thumpd1
Entrez Gene ID
233802
Gene Name
THUMP domain containing 1
Gene Aliases
6330575P11Rik
Chromosome Location
Chr. 7: 119715093 - 119720823 on Build GRCm38
UniGene ID
Mm.26392
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145585.2 NP_663560.1 3 535
GenBank mRNA AK032078.1 - 3 511
AK152912.1 - 3 508
AK183558.1 - 2 255
BC004776.1 - 3 502 3257051

View Full Product Details