Gene Transcripts

Gene Symbol
Prr14
Entrez Gene ID
233895
Gene Name
proline rich 14
Gene Aliases
BC006909
Chromosome Location
Chr. 7: 127470096 - 127476767 on Build GRCm38
UniGene ID
Mm.44183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145589.2 NP_663564.2 7, 8 1673
GenBank mRNA AK133884.1 - 7, 8 1637
AK139559.1 - 8, 9 1448
AK145816.1 - 8, 9 1453
AK151203.1 - 8, 9 1505
AK207496.1 - 30
BC055033.1 - 7, 8 1673 6310272

View Full Product Details