Gene Transcripts

Gene Symbol
Ppan
Entrez Gene ID
235036
Gene Name
peter pan homolog (Drosophila)
Gene Aliases
A230087P06Rik, SSF1
Chromosome Location
Chr. 9: 20888175 - 20892179 on Build GRCm38
UniGene ID
Mm.2930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145610.2 NP_663585.2 5 526
GenBank mRNA AK039027.1 - 5 431
AK085695.1 - 1 1565
BC014688.1 - 5 444 4489442
BC059277.1 - 4 610

Gene Symbol
B130055M24Rik
Entrez Gene ID
654803
Gene Name
RIKEN cDNA B130055M24 gene
Gene Aliases
Chromosome Location
Chr. 7: 127448654 - 127461238 on Build GRCm38
UniGene ID
Mm.423237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK156871.1 - 629

View Full Product Details