Gene Transcripts

Gene Symbol
Rtp3
Entrez Gene ID
235636
Gene Name
receptor transporter protein 3
Gene Aliases
Tmem7
Chromosome Location
Chr. 9: 110984938 - 110990578 on Build GRCm38
UniGene ID
Mm.441138
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ428064.1 - 2 728
AY562227.1 - 2 626
BC112903.1 - 2 728 4196964
BC150678.1 - 2 672 9087589

View Full Product Details