Gene Transcripts

Gene Symbol
Elmo3
Entrez Gene ID
234683
Gene Name
engulfment and cell motility 3
Gene Aliases
9930107J06, BC058752, CED-12, Ced12
Chromosome Location
Chr. 8: 105305399 - 105310623 on Build GRCm38
UniGene ID
Mm.237966
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172760.3 NP_766348.3 20 2854
GenBank mRNA AK037067.1 - 20 2847
BC138887.1 - 20 2811 8861909

View Full Product Details