Gene Transcripts
- Gene Symbol
- Zkscan17
- Entrez Gene ID
- 268417
- Gene Name
- zinc finger with KRAB and SCAN domains 17
- Gene Aliases
- Nizp1, Zfp496, Znf496
- Chromosome Location
- Chr. 11: 59485520 - 59506640 on Build GRCm38
- UniGene ID
- Mm.239793
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY302242.1 |
- |
9 |
2673 |
|
BC040205.1 |
- |
9 |
2677 |
4947290 |
View Full Product Details