Gene Transcripts

Gene Symbol
Kctd8
Entrez Gene ID
243043
Gene Name
potassium channel tetramerisation domain containing 8
Gene Aliases
A730087N02Rik
Chromosome Location
Chr. 5: 69109285 - 69341709 on Build GRCm38
UniGene ID
Mm.234821
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175519.5 NP_780728.3 2 2573
GenBank mRNA AK042569.1 - 2 2429
AK045439.1 - 2 2428
AY615967.1 - 2 2575

View Full Product Details