Gene Transcripts

Gene Symbol
Plch2
Entrez Gene ID
269615
Gene Name
phospholipase C, eta 2
Gene Aliases
A930027K05Rik, PLCeta2, Plc-eta2, Plcl4
Chromosome Location
Chr. 4: 154983115 - 155064091 on Build GRCm38
UniGene ID
Mm.379458
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK044619.1 - 12, 13 1996
AK129147.1 - 12, 13 1757
AY966876.1 - 12, 13 1775

View Full Product Details