Gene Transcripts

Gene Symbol
Glt1d1
Entrez Gene ID
319804
Gene Name
glycosyltransferase 1 domain containing 1
Gene Aliases
5730455A04Rik
Chromosome Location
Chr. 5: 127632130 - 127708750 on Build GRCm38
UniGene ID
Mm.237888
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177005.4 NP_795979.2 2 205
GenBank mRNA AK050379.1 - 2 206
AK077577.1 - 2 206
BC115464.1 - 2 204
BY337475.1 - 2 206

View Full Product Details