Gene Transcripts

Gene Symbol
Sytl5
Entrez Gene ID
236643
Gene Name
synaptotagmin-like 5
Gene Aliases
ENSMUSG00000054453, slp5
Chromosome Location
Chr. X: 9747557 - 9998864 on Build GRCm38
UniGene ID
Mm.246606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290728.1 NP_001277657.1 2 295
NM_177704.3 NP_808372.1 2 295
GenBank mRNA AB098160.1 - 2 294
AB098161.1 - 2 294
BC150158.1 - 2 298 40141504
BC150480.1 - 2 298 40141505

View Full Product Details