Gene Transcripts
- Gene Symbol
- Kif1a
- Entrez Gene ID
- 16560
- Gene Name
- kinesin family member 1A
- Gene Aliases
- ATSV, C630002N23Rik, Gm1626, Kns1
- Chromosome Location
- Chr. 1: 93015455 - 93101890 on Build GRCm38
- UniGene ID
- Mm.276408
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001110315.2 |
NP_001103785.1
|
2, 3 |
277 |
|
NM_001294149.1 |
NP_001281078.1
|
2, 3 |
277 |
|
NM_001294150.1 |
NP_001281079.1
|
1, 2 |
165 |
|
NM_008440.4 |
NP_032466.2
|
2, 3 |
277 |
|
GenBank mRNA |
AK138237.1 |
- |
2, 3 |
219 |
|
AK147640.1 |
- |
2, 3 |
235 |
|
AY426748.1 |
- |
1, 2 |
106 |
|
BC034863.1 |
- |
2, 3 |
222 |
|
BC045161.1 |
- |
2, 3 |
250 |
|
BC054538.1 |
- |
2, 3 |
250 |
|
BC062891.1 |
- |
2, 3 |
256 |
5702372 |
CJ103366.1 |
- |
2, 3 |
278 |
|
CJ115414.1 |
- |
2, 3 |
269 |
|
D29951.1 |
- |
2, 3 |
226 |
|
View Full Product Details