Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008445.2 | NP_032471.2 | 3 | 2563 | |
GenBank mRNA | AF013116.1 | - | 3 | 2139 | |
AK139894.1 | - | 3 | 239 | ||
AK147572.1 | - | 3 | 2365 | ||
AK220283.1 | - | 4 | 2288 | ||
BC052732.1 | - | 3 | 1720 | ||
BC053053.1 | - | 3 | 2505 | ||
BC062648.1 | - | 3 | 2238 | ||
BC127063.1 | - | 3 | 1775 | 40109965 |