Gene Transcripts

Gene Symbol
Klra2
Entrez Gene ID
16633
Gene Name
killer cell lectin-like receptor, subfamily A, member 2
Gene Aliases
Klra30, Ly49b
Chromosome Location
Chr. 6: 131198976 - 131247723 on Build GRCm38
UniGene ID
Mm.4783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170851.1 NP_001164322.1 9 1742
NM_008462.5 NP_032488.4 8 1643
GenBank mRNA BC064711.1 - 9 1643

View Full Product Details