Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008496.4 | NP_032522.2 | 4 | 490 | |
GenBank mRNA | AK003153.1 | - | 4 | 424 | |
AK008773.1 | - | 4 | 424 | ||
AK009151.1 | - | 4 | 433 | ||
BC116838.1 | - | 4 | 451 | 40126157 | |
BC116840.1 | - | 4 | 451 | 40126159 |