Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008910.2 | NP_001008910.1 | 4 | 475 | |
NR_026909.1 | 4 | 740 | |||
XM_011512067.2 | XP_011510369.1 | 4 | 476 | ||
GenBank mRNA | AB020739.1 | - | 3 | 369 | |
AF060798.1 | - | 4 | 446 | ||
AF203910.1 | - | 4 | 442 | ||
AJ005791.1 | - | 3 | 436 | ||
AJ010872.1 | - | 3 | 346 | ||
AK225392.1 | - | 4 | 440 | ||
AK225526.1 | - | 447 | |||
AK292694.1 | - | 4 | 475 | ||
AK302182.1 | - | 4 | 709 | ||
AK310793.1 | - | 4 | 740 | ||
AL552406.3 | - | 4 | 717 | ||
BC002618.2 | - | 4 | 444 | 3143105 | |
BC053998.1 | - | 4 | 440 | 5760866 | |
BT019585.1 | - | 3 | 318 | ||
CR407675.1 | - | 3 | 318 |