Gene Transcripts

Gene Symbol
Smad1
Entrez Gene ID
17125
Gene Name
SMAD family member 1
Gene Aliases
AI528653, Madh1, Madr1, Mlp1, MusMLP
Chromosome Location
Chr. 8: 79338395 - 79399468 on Build GRCm38
UniGene ID
Mm.223717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008539.3 NP_032565.2 7 2074
GenBank mRNA AK017583.1 - 7 2037
AK054104.1 - 7 2035
AK158509.1 - 7 2091
BC058693.1 - 7 2074 6811514
U74359.1 - 7 1941

View Full Product Details