Gene Transcripts

Gene Symbol
Mrc1
Entrez Gene ID
17533
Gene Name
mannose receptor, C type 1
Gene Aliases
AW259686, CD206, MR
Chromosome Location
Chr. 2: 14229414 - 14332023 on Build GRCm38
UniGene ID
Mm.2019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008625.2 NP_032651.2 2, 3 570
GenBank mRNA BC141338.1 - 2, 3 522
Z11974.1 - 2, 3 570

View Full Product Details