Gene Transcripts

Gene Symbol
Cntn3
Entrez Gene ID
18488
Gene Name
contactin 3
Gene Aliases
Pang
Chromosome Location
Chr. 6: 102163304 - 102564620 on Build GRCm38
UniGene ID
Mm.435479
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008779.2 NP_032805.2 11 1541
GenBank mRNA AK140747.1 - 2 130
L01991.1 - 1650

View Full Product Details