Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190978.1 | NP_001177907.1 | 5 | 1274 | |
NM_010623.4 | NP_034753.1 | 5 | 1274 | ||
GenBank mRNA | AB008867.1 | - | 5 | 1272 | |
AK046890.1 | - | 5 | 1027 | ||
AK077868.1 | - | 5 | 1055 |