Gene Transcripts

Gene Symbol
Klf12
Entrez Gene ID
16597
Gene Name
Kruppel-like factor 12
Gene Aliases
2700063E05Rik, AI225908, AI315654, AP-2rep, B130052C06Rik, D530033K05Rik
Chromosome Location
Chr. 14: 99866181 - 100284714 on Build GRCm38
UniGene ID
Mm.396499
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010636.3 NP_034766.2 5 878
GenBank mRNA AB221575.1 - 5 846
BC067408.1 - 5 878 30091275
Y14295.1 - 5 862

View Full Product Details