Gene Transcripts
- Gene Symbol
- Sigmar1
- Entrez Gene ID
- 18391
- Gene Name
- sigma non-opioid intracellular receptor 1
- Gene Aliases
- Oprs1, sigma1R
- Chromosome Location
- Chr. 4: 41738493 - 41741359 on Build GRCm38
- UniGene ID
- Mm.425181
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286538.1 |
NP_001273467.1
|
2, 3 |
470 |
|
NM_001286540.1 |
NP_001273469.1
|
2, 3 |
470 |
|
NM_001286541.1 |
NP_001273470.1
|
1, 2 |
269 |
|
NM_001286551.1 |
NP_001273480.1
|
2, 3 |
423 |
|
NM_001286605.1 |
NP_001273534.1
|
2, 3 |
423 |
|
NM_011014.3 |
NP_035144.1
|
2, 3 |
470 |
|
GenBank mRNA |
AF004927.1 |
- |
2, 3 |
424 |
|
AF030198.1 |
- |
2, 3 |
384 |
|
AK002718.1 |
- |
2, 3 |
403 |
|
AK154300.1 |
- |
2, 3 |
442 |
|
AK159886.1 |
- |
2, 3 |
404 |
|
AY390765.1 |
- |
2, 3 |
363 |
|
AY390766.1 |
- |
2, 3 |
363 |
|
AY390768.1 |
- |
1, 2 |
162 |
|
BC002000.1 |
- |
2, 3 |
419 |
3484605 |
BC019930.1 |
- |
2, 3 |
398 |
5123891 |
BM507398.1 |
- |
2, 3 |
315 |
|
View Full Product Details