Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023418.2 | NP_075907.2 | 3 | 607 | |
GenBank mRNA | AF283667.1 | - | 3 | 519 | |
AK004921.1 | - | 3 | 463 | ||
AK009905.2 | - | 3 | 460 | ||
AK144681.1 | - | 3 | 461 | ||
AK150716.1 | - | 3 | 456 | ||
AK152443.1 | - | 3 | 456 | ||
AK153202.1 | - | 3 | 460 | ||
AK159305.1 | - | 3 | 460 | ||
AK165410.1 | - | 3 | 456 | ||
AK168949.1 | - | 3 | 453 | ||
AK180741.1 | - | 2 | 205 | ||
AK185826.1 | - | 2 | 63 | ||
AK190024.1 | - | 2 | 63 | ||
AK190271.1 | - | 2 | 63 | ||
AK192835.1 | - | 2 | 63 | ||
AK195142.1 | - | 2 | 63 | ||
AK195515.1 | - | 2 | 63 | ||
AK196598.1 | - | 2 | 63 | ||
AK197386.1 | - | 2 | 63 | ||
AK197934.1 | - | 2 | 63 | ||
AK199467.1 | - | 2 | 63 | ||
AK203493.1 | - | 2 | 63 | ||
AK212756.1 | - | 2 | 63 | ||
BC002241.1 | - | 3 | 555 | 3491964 | |
BC005661.1 | - | 3 | 521 | 3588761 | |
BC066844.1 | - | 3 | 444 | 30471755 | |
BC067412.1 | - | 2 | 260 | ||
BC083090.1 | - | 3 | 456 | 30444229 | |
BC106139.1 | - | 3 | 527 | 6528521 |