Gene Transcripts

Gene Symbol
Otx2
Entrez Gene ID
18424
Gene Name
orthodenticle homolog 2
Gene Aliases
E130306E05Rik
Chromosome Location
Chr. 14: 48657452 - 48667644 on Build GRCm38
UniGene ID
Mm.134516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286481.1 NP_001273410.1 4 570
NM_001286482.1 NP_001273411.1 4 546
NM_001286483.1 NP_001273412.1 3 386
NM_144841.4 NP_659090.1 2 468
GenBank mRNA AK053741.1 - 3 383
AK081535.1 - 3 387
AK087527.1 - 3 384
BC017609.1 - 2 468 4527414
BC027104.1 - 4 428 5359966
BC029667.1 - 4 432 5362013
BQ959742.1 - 4 546
CO424557.1 - 4 471

View Full Product Details