Gene Transcripts

Gene Symbol
AKAP17A
Entrez Gene ID
8227
Gene Name
A-kinase anchoring protein 17A
Gene Aliases
721P, AKAP-17A, CCDC133, CXYorf3, DXYS155E, PRKA17A, SFRS17A, XE7, XE7Y
Chromosome Location
Chr. Y: 1591593 - 1602520 on Build GRCh38
UniGene ID
Hs.522572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005088.2 NP_005079.2 2 609
NR_027383.1 2 609
GenBank mRNA BC016935.1 - 2 609
BC028151.1 - 2 577 5217247
BC110496.1 - 2 607 40007662
BC110497.1 - 2 607 40007663
L03426.1 - 2 579
M99578.1 - 2 582

View Full Product Details