Gene Transcripts

Gene Symbol
Epha2
Entrez Gene ID
13836
Gene Name
Eph receptor A2
Gene Aliases
AW545284, Eck, Myk2, Sek-2, Sek2
Chromosome Location
Chr. 4: 141301221 - 141329384 on Build GRCm38
UniGene ID
Mm.2581
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010139.3 NP_034269.2 4 1077
GenBank mRNA AK137704.1 - 4 1059
AK144202.1 - 4 1061
BC140960.1 - 4 1033 9055572
U07634.1 - 4 1038
X78339.1 - 4 1039

View Full Product Details