Gene Transcripts

Gene Symbol
Flt3
Entrez Gene ID
14255
Gene Name
FMS-like tyrosine kinase 3
Gene Aliases
B230315G04, CD135, Flk-2, Flk2, Flt-3, Ly72, wmfl
Chromosome Location
Chr. 5: 147330741 - 147400489 on Build GRCm38
UniGene ID
Mm.194
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010229.2 NP_034359.2 2 382
GenBank mRNA AK045865.1 - 2 205
AK149292.1 - 2 200
AK163640.1 - 2 383
BC109003.1 - 2 175
BC109004.1 - 2 187
M64689.1 - 2 188
X59398.1 - 2 239

View Full Product Details