Gene Transcripts

Gene Symbol
Ifrd2
Entrez Gene ID
15983
Gene Name
interferon-related developmental regulator 2
Gene Aliases
1810034A24Rik, AA470234, AI838527, SKMc15
Chromosome Location
Chr. 9: 107587570 - 107593038 on Build GRCm38
UniGene ID
Mm.215335
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025903.2 NP_080179.1 4, 5 567
GenBank mRNA AK007687.1 - 4, 5 568
AK149496.1 - 4, 5 548
AK165967.1 - 4, 5 561
AK166130.1 - 4, 5 635
AK166258.1 - 4, 5 525
AK178654.1 - 1, 2 38
AY298860.1 - 4, 5 396
BC023486.1 - 4, 5 515
BC037434.1 - 4, 5 523 2651172

View Full Product Details