Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004197.1 | NP_004188.1 | 2 | 414 | |
NM_032454.1 | NP_115830.1 | 2 | 414 | ||
NR_026717.1 | 2 | 599 | |||
GenBank mRNA | AK302111.1 | - | 2 | 613 | |
AK303438.1 | - | 2 | 452 | ||
BC110347.1 | - | 2 | 599 | 5165123 | |
BC121128.1 | - | 1 | 433 | 40120738 | |
BC121129.1 | - | 1 | 432 | 40120740 | |
BC143833.1 | - | 1 | 469 | 9052348 | |
BC143835.1 | - | 1 | 469 | ||
L26260.1 | - | 2 | 414 |