Gene Transcripts

Gene Symbol
Aqp8
Entrez Gene ID
11833
Gene Name
aquaporin 8
Gene Aliases
AI255744, AQP-8
Chromosome Location
Chr. 7: 123462294 - 123468003 on Build GRCm38
UniGene ID
Mm.273175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109045.1 NP_001102515.1 3 478
NM_007474.2 NP_031500.1 3 481
GenBank mRNA AF018952.1 - 3 463
AK146660.1 - 2 160
BC010982.1 - 3 423 4224850
BY713653.1 - 3 483
CF617211.1 - 3 384

View Full Product Details