Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277216.1 | NP_001264145.1 | 8 | 800 | |
NM_001277217.1 | NP_001264146.1 | 8 | 826 | ||
NM_001277218.1 | NP_001264147.1 | 7 | 839 | ||
NM_001277220.1 | NP_001264149.1 | 8 | 974 | ||
NM_007560.4 | NP_031586.1 | 7 | 993 | ||
GenBank mRNA | AK086130.1 | - | 8 | 786 | |
AK140158.1 | - | 8 | 898 | ||
AK162844.1 | - | 7 | 674 | ||
BC065106.1 | - | 7 | 774 | 30357130 | |
BC065143.1 | - | 7 | 980 | 30629345 | |
BC138638.1 | - | 7 | 720 | 8861658 | |
BC138640.1 | - | 7 | 720 | 8861660 | |
Z23143.1 | - | 8 | 818 |