Gene Transcripts

Gene Symbol
Csn1s2a
Entrez Gene ID
12993
Gene Name
casein alpha s2-like A
Gene Aliases
Csng, PP22
Chromosome Location
Chr. 5: 87774567 - 87788797 on Build GRCm38
UniGene ID
Mm.474059
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK142688.1 - 14 827
AK164738.1 - 14 831
AK164778.1 - 1 29
BB661763.1 - 1 25
D10215.1 - 14 832

Gene Symbol
Gm33867
Entrez Gene ID
102636929
Gene Name
predicted gene, 33867
Gene Aliases
Chromosome Location
Chr. 3: 143743121 - 143772786 on Build GRCm38
UniGene ID
Mm.317208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC032178.1 - 2027

View Full Product Details