Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198872.1 | NP_001185801.1 | 3 | 354 | |
NM_001198873.1 | NP_001185802.1 | 3 | 354 | ||
NM_001198874.1 | NP_001185803.1 | 3 | 354 | ||
NM_001198875.1 | NP_001185804.1 | 3 | 354 | ||
NM_001198876.1 | NP_001185805.1 | 3 | 354 | ||
NM_001198877.1 | NP_001185806.1 | 3 | 343 | ||
NM_001198878.1 | NP_001185807.1 | 3 | 343 | ||
NM_010064.4 | NP_034194.1 | 3 | 343 | ||
GenBank mRNA | AF063231.1 | - | 3 | 282 | |
AK088015.1 | - | 3 | 314 | ||
AK135477.1 | - | 3 | 285 | ||
AK141837.1 | - | 3 | 322 | ||
AK150173.1 | - | 3 | 304 | ||
AK164322.1 | - | 3 | 278 | ||
AK164437.1 | - | 3 | 278 | ||
AK168730.1 | - | 3 | 318 | ||
BC139374.1 | - | 3 | 238 | 8862396 | |
BC139375.1 | - | 3 | 238 | 8862397 | |
BY065048.1 | - | 3 | 357 | ||
BY226221.1 | - | 3 | 344 | ||
GU992208.1 | - | 3 | 245 | ||
GU992209.1 | - | 3 | 245 | ||
GU992210.1 | - | 3 | 245 | ||
GU992211.1 | - | 3 | 283 | ||
GU992212.1 | - | 3 | 283 |