Gene Transcripts

Gene Symbol
Acadm
Entrez Gene ID
11364
Gene Name
acyl-Coenzyme A dehydrogenase, medium chain
Gene Aliases
AU018656, MCAD
Chromosome Location
Chr. 3: 153922353 - 153944643 on Build GRCm38
UniGene ID
Mm.10530
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007382.5 NP_031408.1 8 878
GenBank mRNA AK131793.1 - 8 680
AK146444.1 - 8 683
BC013498.1 - 8 663 4222450
U07159.1 - 8 667

View Full Product Details