Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008904.2 | NP_032930.1 | 4 | 626 | |
GenBank mRNA | AB061324.1 | - | 4 | 549 | |
AF049330.1 | - | 4 | 578 | ||
AK138668.1 | - | 4 | 627 | ||
AK143753.1 | - | 4 | 578 | ||
BC066868.1 | - | 4 | 521 | 30094033 | |
JX866948.1 | - | 4 | 514 |