Gene Transcripts

Gene Symbol
Cept1
Entrez Gene ID
99712
Gene Name
choline/ethanolaminephosphotransferase 1
Gene Aliases
9930118K05Rik, mCEPT1
Chromosome Location
Chr. 3: 106502260 - 106547802 on Build GRCm38
UniGene ID
Mm.486418
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293694.1 NP_001280623.1 2 401
NM_133869.4 NP_598630.2 2 401
GenBank mRNA AK037116.1 - 2 361
BC021753.1 - 2 330 4167222
BC023783.1 - 2 341 5346896

View Full Product Details