Gene Transcripts
- Gene Symbol
- Dpp9
- Entrez Gene ID
- 224897
- Gene Name
- dipeptidylpeptidase 9
- Gene Aliases
- 6430584G11Rik, A330078I11, DPP IX, DPRP2
- Chromosome Location
- Chr. 17: 56186674 - 56218889 on Build GRCm38
- UniGene ID
- Mm.205535
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172624.3 |
NP_766212.2
|
21 |
3433 |
|
GenBank mRNA |
AK078301.1 |
- |
21 |
3381 |
|
View Full Product Details