Gene Transcripts

Gene Symbol
Lrsam1
Entrez Gene ID
227738
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
Chromosome Location
Chr. 2: 32925215 - 32961654 on Build GRCm38
UniGene ID
Mm.248037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199302.2 NP_955006.1 25 3713
GenBank mRNA BC049146.1 - 25 3709
CJ236327.1 - 1 151

View Full Product Details