Gene Transcripts

Gene Symbol
Pck2
Entrez Gene ID
74551
Gene Name
phosphoenolpyruvate carboxykinase 2 (mitochondrial)
Gene Aliases
1810010O14Rik, 9130022B02Rik, PEPCK-M
Chromosome Location
Chr. 14: 55540263 - 55550777 on Build GRCm38
UniGene ID
Mm.29856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028994.2 NP_083270.1 3 547
GenBank mRNA AK030501.1 - 3 548
AK031612.1 - 3 392
AK034927.1 - 3 426
AK147972.1 - 3 401
AK165094.1 - 3 425
AK170242.1 - 3 401
BC010318.1 - 3 410 3596227
BC011126.1 - 3 407
BC023437.1 - 1 15

View Full Product Details