Gene Transcripts

Gene Symbol
Rnase6
Entrez Gene ID
78416
Gene Name
ribonuclease, RNase A family, 6
Gene Aliases
9530043P15Rik, BB119466
Chromosome Location
Chr. 14: 51123913 - 51132187 on Build GRCm38
UniGene ID
Mm.103835
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030098.2 NP_084374.2 2 876
GenBank mRNA AK020595.1 - 2 819
AK038708.1 - 3 1192
AY545655.1 - 2 854
BC094892.1 - 2 831 6432787

View Full Product Details