Gene Transcripts
- Gene Symbol
- ACSM3
- Entrez Gene ID
- 6296
- Gene Name
- acyl-CoA synthetase medium-chain family member 3
- Gene Aliases
- SA, SAH
- Chromosome Location
- Chr. 16: 20763990 - 20797157 on Build GRCh38
- UniGene ID
- Hs.706754
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005622.3 |
NP_005613.2
|
2, 3 |
408 |
|
NM_202000.2 |
NP_973729.1
|
2, 3 |
408 |
|
XM_011545911.1 |
XP_011544213.1
|
2, 3 |
402 |
|
XM_017023523.1 |
XP_016879012.1
|
2, 3 |
402 |
|
XM_017023524.1 |
XP_016879013.1
|
1, 2 |
272 |
|
GenBank mRNA |
AK223139.1 |
- |
2, 3 |
377 |
|
AK290322.1 |
- |
2, 3 |
377 |
|
AK296122.1 |
- |
1, 2 |
96 |
|
BC002790.2 |
- |
1, 2 |
267 |
3617826 |
BI770558.1 |
- |
2, 3 |
408 |
|
D16350.1 |
- |
1, 2 |
248 |
|
X80062.1 |
- |
2, 3 |
357 |
|
View Full Product Details