Gene Transcripts

Gene Symbol
Slc25a32
Entrez Gene ID
69906
Gene Name
solute carrier family 25, member 32
Gene Aliases
2610043O12Rik, Mftc
Chromosome Location
Chr. 15: 39094191 - 39112768 on Build GRCm38
UniGene ID
Mm.17875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172402.3 NP_765990.2 3 580
GenBank mRNA AK031187.1 - 3 385
AK145178.1 - 3 540
AK167501.1 - 3 390
AK170643.1 - 3 549
AK172029.1 - 3 548
AK172597.1 - 3 538
AK187971.1 - 2 90
BB641480.1 - 3 581
BC031874.1 - 3 514 5149984

View Full Product Details