Gene Transcripts

Gene Symbol
Clec18a
Entrez Gene ID
353287
Gene Name
C-type lectin domain family 18, member A
Gene Aliases
Mrcl
Chromosome Location
Chr. 8: 111069496 - 111092426 on Build GRCm38
UniGene ID
Mm.135495
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181549.3 NP_853527.2 3 445
GenBank mRNA AK132888.1 - 5 1252
AY223868.1 - 653
AY223869.1 - 624
AY223870.1 - 628
AY223871.1 - 534
AY223872.1 - 599
AY223873.1 - 505

View Full Product Details