Gene Transcripts

Gene Symbol
Scyl2
Entrez Gene ID
213326
Gene Name
SCY1-like 2 (S. cerevisiae)
Gene Aliases
AU015688, AW536445, BC030932, CVAK104, D10Ertd802e
Chromosome Location
Chr. 10: 89638720 - 89686292 on Build GRCm38
UniGene ID
Mm.27651
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198021.2 NP_932138.1 3 578
GenBank mRNA AK139761.1 - 3 562
AK167276.1 - 3 562
BC042443.1 - 3 578 4037878

View Full Product Details