Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164661.1 | NP_001158133.1 | 9 | 1054 | |
NM_001164662.1 | NP_001158134.1 | 8 | 1001 | ||
NM_011370.3 | NP_035500.2 | 8 | 1001 | ||
GenBank mRNA | AF072697.1 | - | 8 | 796 | |
AJ567911.1 | - | 7 | 763 | ||
AK044175.1 | - | 8 | 823 | ||
AK044651.1 | - | 8 | 805 | ||
AK128952.1 | - | 8 | 788 | ||
AK146613.1 | - | 8 | 804 | ||
AK152558.1 | - | 9 | 870 | ||
AK157773.1 | - | 8 | 789 | ||
AK167068.1 | - | 8 | 824 | ||
AK220320.1 | - | 8 | 842 | ||
BC047135.1 | - | 8 | 790 | 6308554 | |
BC052713.1 | - | 8 | 866 | 6835403 | |
BC054429.1 | - | 7 | 774 | 5345535 |