Gene Transcripts

Gene Symbol
Clcnka
Entrez Gene ID
12733
Gene Name
chloride channel Ka
Gene Aliases
C75963, CLC-K1, Clcnk1
Chromosome Location
Chr. 4: 141384611 - 141398706 on Build GRCm38
UniGene ID
Mm.167900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146307.1 NP_001139779.1 19 2263
NM_024412.3 NP_077723.3 20 2267
GenBank mRNA BB743618.1 - 2 297
BC037077.1 - 20 2271 5042736

View Full Product Details