Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172068.1 | NP_001165539.1 | 4 | 690 | |
NM_001172069.1 | NP_001165540.1 | 3 | 599 | ||
NM_001172070.1 | NP_001165541.1 | 3 | 580 | ||
NM_001172071.1 | NP_001165542.1 | 3 | 597 | ||
NM_027838.3 | NP_082114.1 | 2 | 487 | ||
GenBank mRNA | AK018606.1 | - | 3 | 578 | |
AK029856.1 | - | 2 | 477 | ||
AK049120.1 | - | 4 | 680 | ||
AK136214.1 | - | 3 | 570 | ||
BC037443.1 | - | 2 | 459 | 3599077 | |
CO431667.1 | - | 3 | 564 | ||
DV645349.1 | - | 2 | 487 |