Gene Transcripts

Gene Symbol
Papd4
Entrez Gene ID
100715
Gene Name
PAP associated domain containing 4
Gene Aliases
8030446C20Rik, AI649009, GLD-2, Gld2
Chromosome Location
Chr. 13: 93146275 - 93192970 on Build GRCm38
UniGene ID
Mm.242865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133905.2 NP_598666.1 5 709
GenBank mRNA AK033141.1 - 2 46
AK146086.1 - 5 693
AM419011.1 - 4 482
BC016629.1 - 5 709 3595512

View Full Product Details