Gene Transcripts

Gene Symbol
Csnk1g3
Entrez Gene ID
70425
Gene Name
casein kinase 1, gamma 3
Gene Aliases
3300002K07Rik, C330049O21Rik
Chromosome Location
Chr. 18: 53862082 - 53955684 on Build GRCm38
UniGene ID
Mm.368668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152809.2 NP_690022.2 8 1547
GenBank mRNA AK032675.1 - 2 102
BC033601.1 - 5 562
BQ443928.1 - 5 581

View Full Product Details